Learn More
Kingfisher Biotech Inc Mouse VEGF-C Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1806M005
Encompass
The Mouse/Rat/Hamster VEGF-C yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse/Rat/Hamster VEGF-C applications are for cell culture, ELISA standard, and Western Blot Control. Mouse/Rat/Hamster VEGF-C yeast-derived recombinant protein can be purchased in multiple sizes. Mouse/Rat/Hamster VEGF-C Specifications: (Molecular Weight: 13.1 kDa) (Amino Acid Sequence: AAHYNTEILK SIDNEWRKTQ CMPREVCIDV GKEFGAATNT FFKPPCVSVY RCGGCCNSEG LQCMNTSTGY LSKTLFEITV PLSQGPKPVT ISFANHTSCR CMSKLDVYRQ VHSIIRR (117)) (Gene ID: 22341). For research use only. Made in the USA
Catalog No. 50-253-2742