Learn More
Alpco Ltd Mouse / Rat IGFBP-2 ELISA
SDP
Supplier:  Alpco Ltd 22BP2MSE01
Encompass
The Mouse / Rat IGFBP-2 ELISA is for the quantitative determination of IGFBP-2 in rat and mouse serum. For Research Use only. Not For In Vitro Diagnostic Use.
Dynamic range
0.01-200 ng/mL
Sensitivity
0.01 ng/mL
Sample size
10 µL
Catalog No. NC1653307