Learn More
Euthanex Sure-Seal Mouse/Rat Chamber, 9"" L x 4"" W x 4"" H
Catalog No. 501540171
Encompass_Preferred
Sure-Seal Mouse/Rat Chamber, 9" L x 4" W x 4" H, heavy-duty acrylic, gasket seal, locking latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Accommodates oxygen-flush before transferring animals out of chamber.
Catalog No. 50-154-0171 Supplier Euthanex Supplier No. EZ178
Sure-Seal Mouse/Rat Chamber, 9" L x 4" W x 4" H, heavy-duty acrylic, gasket seal, locking latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Accommodates oxygen-flush before transferring animals out of chamber.