Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat RGM-A Quantikine ELISA Kit
SDP

Catalog No. MRGMA0
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MRGMA0 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MRGMA0 Supplier R&D Systems™ Supplier No. MRGMA0

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

a 4.5 hour solid phase ELISA designed to measure mouse and rat RGM-A in cell culture supernates, tissue homogenates, serum, and plasma.

The Mouse/Rat RGM-A Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat RGM-A in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul).

Specifications

Assay Range 0.08 - 5ng/mL
Assay Sensitivity 0.019ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse and Rat RGM-A
For Use With (Application) ELISA
Gene ID (Entrez) 244058
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.