Learn More
Abcam Mouse/Rat Thyroxine (T4) ELISA Kit, 96TEST
Supplier:  Abcam AB28525896TEST
Encompass_Preferred
Mouse/Rat Thyroxine (T4) ELISA Kit ab285258 is used to quantify Mouse/Rat Thyroxine (T4) with a sensitivity of 55.8 pg/ml. Competitive ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5502