Learn More
Abcam Mouse/Rat Triiodothyronine (T3) ELISA Kit, 96TEST
Supplier:  Abcam AB28525996TEST
Encompass_Preferred
Mouse/Rat Triiodothyronine (T3) ELISA Kit is a Competitive ELISA kit for the measurement of Mouse/Rat Triiodothyronine (T3) in Rat, Mouse in Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5455