Learn More
Sino Biological Anti-RBP4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50170RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse RBP4 (rM RBP4; Catalog#50170-M08H; NP_035385.1; Met 1-Leu 201). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0670