Learn More
Cell Signaling Technology Mouse Reactive PANoptosis Antibody Sampler Kit, 1 Kit
SDP
Catalog No. NC2229295
Encompass
The Mouse Reactive PANoptosis Antibody Sampler Kit provides an economical means of detecting the activation of PANoptosis in mouse samples. This kit enables the simultaneous analysis of multiple key proteins involved in the coordinated cell death pathway known as PANoptosis, which integrates features of apoptosis, pyroptosis, and necroptosis. The kit includes sufficient primary antibody volume to perform two western blot experiments for each target.
  • Validated for western blotting, immunofluorescence, flow cytometry, immunoprecipitation, and immunohistochemistry.
  • Includes primary antibodies for RIPK3, phospho-RIPK3, IL-1β, cleaved IL-1β, Caspase-3, MLKL, phospho-MLKL, Gasdermin D, and cleaved Gasdermin D.
  • Contains HRP-linked secondary antibody for convenient detection.
  • Designed for mouse-reactive studies.
This item is not returnable. View return policy
Catalog No. NC2229295 Supplier Cell Signaling Technology Supplier No. 70934T
The Mouse Reactive PANoptosis Antibody Sampler Kit provides an economical means of detecting the activation of PANoptosis in mouse samples. This kit enables the simultaneous analysis of multiple key proteins involved in the coordinated cell death pathway known as PANoptosis, which integrates features of apoptosis, pyroptosis, and necroptosis. The kit includes sufficient primary antibody volume to perform two western blot experiments for each target.
  • Validated for western blotting, immunofluorescence, flow cytometry, immunoprecipitation, and immunohistochemistry.
  • Includes primary antibodies for RIPK3, phospho-RIPK3, IL-1β, cleaved IL-1β, Caspase-3, MLKL, phospho-MLKL, Gasdermin D, and cleaved Gasdermin D.
  • Contains HRP-linked secondary antibody for convenient detection.
  • Designed for mouse-reactive studies.