Learn More
Cell Signaling Technology Exosome Marker Antibody Sampler Kit, Mouse Reactive, 1 Kit
SDP
Catalog No. 502729774
Encompass
The Mouse Reactive Exosome Marker Antibody Sampler Kit provides a cost-effective solution for detecting protein markers commonly associated with exosomes. This kit includes primary antibodies validated for analyzing exosomal protein profiles via western blot and other immunological assays. The selection of markers targets integral membrane proteins and components of the endosomal sorting complex required for transport (ESCRT) pathway to facilitate the study of exosome biogenesis and composition.
  • Includes antibodies for flotillin-1, CD81, HSP70, Alix, CD9, and TSG101
  • Provides sufficient reagent to perform two western blot experiments for each primary antibody
  • Supplied with an HRP-linked anti-rabbit IgG secondary antibody
  • Contains antibodies optimized for specific reactivity with mouse samples
Catalog No. 50-272-9774 Supplier Cell Signaling Technology Supplier No. 96128T
The Mouse Reactive Exosome Marker Antibody Sampler Kit provides a cost-effective solution for detecting protein markers commonly associated with exosomes. This kit includes primary antibodies validated for analyzing exosomal protein profiles via western blot and other immunological assays. The selection of markers targets integral membrane proteins and components of the endosomal sorting complex required for transport (ESCRT) pathway to facilitate the study of exosome biogenesis and composition.
  • Includes antibodies for flotillin-1, CD81, HSP70, Alix, CD9, and TSG101
  • Provides sufficient reagent to perform two western blot experiments for each primary antibody
  • Supplied with an HRP-linked anti-rabbit IgG secondary antibody
  • Contains antibodies optimized for specific reactivity with mouse samples