Learn More
Cell Signaling Technology M1 and M2 Macrophage IHC Antibody Sampler Kit, Mouse Reactive, 1 Kit
SDP
Catalog No. NC2055466
Encompass
The Mouse Reactive M1 vs M2 Macrophage IHC Antibody Sampler Kit provides an economical means of characterizing the extent of M1 and M2 macrophages in formalin-fixed, paraffin-embedded tissue samples. This kit includes a selection of antibodies against general macrophage markers, as well as specific markers for the classically activated M1 and alternatively activated M2 phenotypes. An HRP-linked secondary antibody is provided for signal detection in immunohistochemistry applications.
  • Economical selection of antibodies for macrophage characterization
  • Validated for immunohistochemistry on paraffin-embedded tissue samples
  • Includes general macrophage markers as well as M1 and M2 polarization markers
  • Supplied with HRP-linked secondary antibody for convenient detection
  • Provided in a stable storage buffer containing 50% glycerol
Catalog No. NC2055466 Supplier Cell Signaling Technology Supplier No. 97624T
The Mouse Reactive M1 vs M2 Macrophage IHC Antibody Sampler Kit provides an economical means of characterizing the extent of M1 and M2 macrophages in formalin-fixed, paraffin-embedded tissue samples. This kit includes a selection of antibodies against general macrophage markers, as well as specific markers for the classically activated M1 and alternatively activated M2 phenotypes. An HRP-linked secondary antibody is provided for signal detection in immunohistochemistry applications.
  • Economical selection of antibodies for macrophage characterization
  • Validated for immunohistochemistry on paraffin-embedded tissue samples
  • Includes general macrophage markers as well as M1 and M2 polarization markers
  • Supplied with HRP-linked secondary antibody for convenient detection
  • Provided in a stable storage buffer containing 50% glycerol