Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Mouse Reactive Pyroptosis Antibody Sampler Kit, 1 Kit
SDP

Catalog No. 502051382
Encompass

The Mouse Reactive Pyroptosis Antibody Sampler Kit provides an economical means of detecting proteins that are used as readouts for pyroptosis. The kit includes enough antibodies to perform two western blot experiments with each primary antibody.

  • Contains nine primary antibodies specifically selected for mouse reactivity in pyroptosis research.
  • Includes secondary antibody for detection.
  • Validated for multiple applications including western blotting, immunofluorescence, immunohistochemistry (paraffin), flow cytometry, and immunoprecipitation.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol and less than 0.02% sodium azide.
  • Store at −20°C.

Catalog No. 50-205-1382 Supplier Cell Signaling Technology Supplier No. 98303T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Mouse Reactive Pyroptosis Antibody Sampler Kit provides an economical means of detecting proteins that are used as readouts for pyroptosis. The kit includes enough antibodies to perform two western blot experiments with each primary antibody.

  • Contains nine primary antibodies specifically selected for mouse reactivity in pyroptosis research.
  • Includes secondary antibody for detection.
  • Validated for multiple applications including western blotting, immunofluorescence, immunohistochemistry (paraffin), flow cytometry, and immunoprecipitation.
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/ml BSA, 50% glycerol and less than 0.02% sodium azide.
  • Store at −20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.