Learn More
Biomatik Corporation Mouse Receptor For Advanced Glycation Endproducts (RAGE) CLIA Kit
Supplier:  Biomatik Corporation EKU08906
Encompass_Preferred
Mouse Receptor For Advanced Glycation Endproducts (RAGE) CLIA Kit for detecting RAGE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q62151. Gene ID: 11596. Target Synonyms: AGER; Receptor For Advanced Glycation Endproducts. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8596