Learn More
American Research Products Inc Mouse Receptor interacting ser
Catalog No. 5000327390
Encompass
Mouse Receptor-interacting serine/threonine-protein kinase 3 (RIPK3) ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27390 Supplier American Research Products Inc Supplier No. CSBEL019737MO
Add to Cart
Add to Cart
Mouse Receptor-interacting serine/threonine-protein kinase 3 (RIPK3) ELISA kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich