Learn More
Biomatik Corporation Mouse Receptor Interacting Serine Threonine Kinase 1 (RIPK1) ELISA Kit
Supplier:  Biomatik Corporation EKU12290
Encompass_Preferred
Mouse Receptor Interacting Serine Threonine Kinase 1 (RIPK1) ELISA Kit for detecting RIPK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60855. Gene ID: 19766. Target Synonyms: RIP; Receptor-Interacting Protein; Cell death protein RIP; Serine/threonine-protein kinase RIP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5682