Learn More
STEMCELL Technologies Mouse Recombinant GM-CSF (E. coli-expressed), Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78017
Encompass
Use granulocyte-macrophage colony-stimulating factor (GM-CSF) to induce the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages (Barreda et al. Dev Comp Immunol, 2004). GM-CSF can stimulate the development of DCs that ingest, process, and present antigens to the immune system (Francisco-Cruz et al. Med Oncol, 2014). It is produced by multiple cell types, including macrophages, dendritic cells, endothelial cells, and fibroblasts. Together with other cytokines such as stem cell factor, IL-3, erythropoietin, and thrombopoietin, GM-CSF also stimulates erythroid and megakaryocyte progenitor cells (Barreda et al.). Mouse recombinant GM-CSF (E.coli-expressed) from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 50 pg/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: Colony-stimulating factor 2, CSF-2, MGI-1GM, Pluripoitin-alpha. Visit STEMCELL.com for more information.
Catalog No. 50-197-6277
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov