Learn More
STEMCELL Technologies Mouse Recombinant IL-2, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 78081.1
Encompass
Use interleukin 2 (IL-2) to modulate cellular proliferation and survival through JAK3, STAT5, and AKT pathways (Ma et al. Annu Rev Immunol, 2006). IL-2 is a monomeric cytokine that binds to heterotrimeric receptors consisting of CD25, CD122, and CD132. It downregulates immune responses during thymic development, influences the development of CD4+CD25+ regulatory T cells, and affects development of follicular helper T cells. IL-2 controls inflammation by inhibiting Th17 differentiation (Banchereau et al. Nat Immunol, 2012). Targeted deletions of the IL-2 gene in mice resulted in development of autoimmune hemolytic anemia, followed by ulcerative colitis. Similar effects were observed in mice deficient in IL-2 receptor α (Gaffen & Liu. Cytokine, 2004). Mouse recombinant IL-2 from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 5 ng/mL; endotoxin levels verified ≤ 1 EU/μg protein. Alternative names: Aldesleukin, Interleukin-2, T cell growth factor, TCGF.
Catalog No. 50-197-6422
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov