Learn More
STEMCELL Technologies Mouse Recombinant IL-7, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 78054
Encompass
Interleukin 7 (IL-7) is a member of the type I cytokine family that is critical for T and B cell development and survival. It is produced by non-hematopoietic cells in the thymus, lymphoid organs, and by bone marrow stromal cells (Lundström et al.). IL-7 binds to a receptor composed of common gamma chain and IL-7Ra (CD127) and signals through the JAK/STAT and PI3K pathways. IL-7 regulates the survival of naïve and memory CD4+ and CD8+ T cells, γδ T cells, NK T cells, innate lymphoid cells, and T regulatory cells (Carrette & Surh), and provides signals to promote survival, proliferation, and differentiation during B cell development (Clark et al.; Corfe & Paige).
Catalog No. 50-197-6363