Learn More
Sino Biological Anti-Reg3A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50185RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse REG3A (rM REG3A; Catalog#50185-M08H; NP_035389.1; Met1-Gln175). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3385