Learn More
Sino Biological Anti-Reg3A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50185T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse REG3A (rM REG3A; Catalog#50185-M08H; NP_035389.1; Met1-Gln175). REG3A specific IgG was purified by Mouse REG3A affinity chromatography.
Catalog No. 50-255-3384