Learn More
Sino Biological Recombinant Anti-Reg3b Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51153R065100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse REG3B (rM REG3B; Catalog#51153-M08H; NP_035166.1; Met1-Gly175).
Catalog No. 50-255-2978