Learn More
Sino Biological Anti-Reg3b Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51153T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse REG3B (rM REG3B; Catalog#51153-M08H; NP_035166.1; Met1-Gly175). REG3B specific IgG was purified by Mouse REG3B affinity chromatography.
Catalog No. 50-255-2977