Learn More
Sino Biological Mouse REG3D Protein 100ug (Sino Biological)
Supplier:  Sino Biological 50540M08H100
Encompass_Preferred
A DNA sequence encoding the mouse REG3D isoform 1 (NP_038921.2) extracellular domain (Met 1-Gly 175) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2959