Learn More
Biomatik Corporation Mouse Reg3β (RegeneRating islet-derived protein 3-beta) ELISA Kit
Supplier:  Biomatik Corporation EKF58415
Encompass_Preferred
Mouse Reg3β (RegeneRating islet-derived protein 3-beta) ELISA Kit for detecting Reg3β in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35230. Target Synonyms: Reg3β; Reg3b; HIP; PAP1; Pap; REG-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5986