Learn More
American Research Products Inc Mouse Regenerating islet deriV
Catalog No. 5000327776
Encompass
Mouse Regenerating islet-deriVed protein 3-gamma (REG3G) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27776 Supplier American Research Products Inc Supplier No. CSBEL019549MO
Mouse Regenerating islet-deriVed protein 3-gamma (REG3G) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich