Learn More
American Research Products Inc Mouse Regenerating islet deriV
Catalog No. 5000328217
Encompass
Mouse Regenerating islet-deriVed protein 3-alpha (REG3A) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28217 Supplier American Research Products Inc Supplier No. CSBEL019548MO
Mouse Regenerating islet-deriVed protein 3-alpha (REG3A) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich