Learn More
Biomatik Corporation Mouse Regucalcin (RGN) ELISA Kit
Supplier:  Biomatik Corporation EKU11391
Encompass_Preferred
Mouse Regucalcin (RGN) ELISA Kit for detecting RGN in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64374. Gene ID: 19733. Target Synonyms: RC; SMP30; GNL; Senescence Marker Protein-30; Gluconolactonase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8151