Learn More
American Research Products Inc Mouse regulated on actiVation
Catalog No. 5000329307
Encompass
Mouse regulated on actiVation in normal T-cell eXpressed and secreted RANTES ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29307 Supplier American Research Products Inc Supplier No. CSBE09256M
Mouse regulated on actiVation in normal T-cell eXpressed and secreted RANTES ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich