Learn More
American Research Products Inc Mouse regulated on actiVation
Supplier:  American Research Products Inc CSBE09256MIS
Encompass
Mouse regulated on actiVation in normal T-cell eXpressed and secreted RANTES ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29969