Learn More
Sino Biological Mouse REN MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50279R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse REN1 (rM REN1; Catalog#50279-M08H; NP_112469.1; Met 1-Arg 402).
Catalog No. 50-237-2670