Learn More
Sino Biological Anti-Renin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50279RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse REN1 (rM REN1; Catalog#50279-M08H; NP_112469.1; Met 1-Arg 402). REN1 specific IgG was purified by mouse REN1 affinity chromatography.
Catalog No. 50-255-3316