Learn More
Raybiotech Inc MOUSE RENIN 1 ELISA
Supplier:  Raybiotech Inc ELMRENIN11
Encompass_Preferred
RayBio Mouse Renin 1 ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 6 pg/ml - 50000 pg/ml
Catalog No. 50-104-4785