Learn More
Biomatik Corporation Mouse Resistin (RETN) CLIA Kit
Supplier:  Biomatik Corporation EKU08921
Encompass_Preferred
Mouse Resistin (RETN) CLIA Kit for detecting RETN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 54.9-40,000pg/ml. Sensitivity: 18.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q99P87. Gene ID: 57264. Target Synonyms: ADSF; FIZZ3; RETN1; RSTN; XCP1; Adipose tissue-specific secretory factor; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; Cysteine-rich secreted FIZZ3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7071