Learn More
Braintree Scientific Mouse Tail Illuminator Restrainer,Tube Only-Small 1.12""""id
SDP
Catalog No. NC9528078
Encompass
The Mouse Tail Illuminator efficiently illuminates and warms a mouse tail for easy injection or bleeding. Each unit comes with a restraining tube and illumination source. Additional tubes are available. Restraining tube slides off base for easy cleaning. Illuminated slot for easy tail vein injection, etc. Sliding doors keep mouse gently restrained. **Sold for research purposes only and not for human use**
Catalog No. NC9528078 Supplier Braintree Scientific Supplier No. MTITUBE2
The Mouse Tail Illuminator efficiently illuminates and warms a mouse tail for easy injection or bleeding. Each unit comes with a restraining tube and illumination source. Additional tubes are available. Restraining tube slides off base for easy cleaning. Illuminated slot for easy tail vein injection, etc. Sliding doors keep mouse gently restrained. **Sold for research purposes only and not for human use**