Learn More
SAI Infusion Technologies Solid Brass "Cozy" Mouse Restrainer
SDP
Catalog No. NC0249573
Encompass
SAI's Cozy Mouse Restrainer restrains mice for tail vein injections or sampling. Solid brass construction makes a heavy and stable base during procedures, and retains heat for long periods to improve animal comfort and vasodilation of the tail. Product is for laboratory animal research only.
Catalog No. NC0249573 Supplier SAI Infusion Technologies Supplier No. MR01
SAI's Cozy Mouse Restrainer restrains mice for tail vein injections or sampling. Solid brass construction makes a heavy and stable base during procedures, and retains heat for long periods to improve animal comfort and vasodilation of the tail. Product is for laboratory animal research only.