Learn More
IBI SCIENTIFIC 3P Mouse Restraint Stand
Catalog No. NC1862806
Encompass_Preferred
Holds mouse restraint at 45 degree angle. Allows for easy access to tail for bleeds. Holds restraint securely. The MH-550 is designed to hold the MH-100 and MH-125 at a 45 degree angle to allow for easy access to the mouse tail for bleeds or other procedures. The stand is constructed of cast acrylic for years of reliable service.
Catalog No. NC1862806 Supplier IBI SCIENTIFIC 3P Supplier No. MH100
Holds mouse restraint at 45 degree angle. Allows for easy access to tail for bleeds. Holds restraint securely. The MH-550 is designed to hold the MH-100 and MH-125 at a 45 degree angle to allow for easy access to the mouse tail for bleeds or other procedures. The stand is constructed of cast acrylic for years of reliable service.