Learn More
Biomatik Corporation Mouse Reticulocalbin 2 (RCN2) ELISA Kit
Supplier:  Biomatik Corporation EKU07090
Encompass_Preferred
Mouse Reticulocalbin 2 (RCN2) ELISA Kit for detecting RCN2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BP92. Gene ID: 26611. Target Synonyms: E6BP; ERC-55; ERC55; Endoplasmic Reticulum Calcium-Binding Protein 55kD; E6-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8779