Learn More
American Research Products Inc Mouse Retinol binding protein
Catalog No. 5000326767
Encompass
Mouse Retinol-binding protein RBP ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma urine tissue homogenates Method Competitive
Catalog No. 50-003-26767 Supplier American Research Products Inc Supplier No. CSBE09151M
Mouse Retinol-binding protein RBP ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma urine tissue homogenates Method Competitive