Learn More
Sino Biological Recombinant Anti-Resistin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50488R020100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Resistin (rM Resistin; Q99P87; Ser21-Ser114).
Catalog No. 50-255-3247