Learn More
BIOHIPPO Mouse RGS5 Microsample
SDP
Catalog No. 5000315161
Encompass
Mouse RGS5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RGS5 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 072 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse RGS5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse RGS5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse RGS5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15161 Supplier BIOHIPPO Supplier No. BHE15210864
Mouse RGS5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RGS5 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 072 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse RGS5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse RGS5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse RGS5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color