Learn More
Biomatik Corporation Mouse Ribonuclease T2 (RNASET2) CLIA Kit
Supplier:  Biomatik Corporation EKU09756
Encompass_Preferred
Mouse Ribonuclease T2 (RNASET2) CLIA Kit for detecting RNASET2 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: C0HKG5. Gene ID: 100037283. Target Synonyms: Rnase-T2; RNASE6PL; RNASE6-PL; Ribonuclease 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5198