Learn More
ACROBiosystems FITC-Labeled Anti-Rituximab Antibody, Mouse IgG1
Catalog No. 502718975
Encompass_Preferred
Rituxan is a genetically engineered chimeric murine/human monoclonal antibody directed against the CD20 antigen found on the surface of normal and malignant B lymphocytes. The antibody is an IgG1 kappa immunoglobulin containing murine light- and heavy-chain variable region sequences and human constant region sequences. Rituximab is composed of two heavy chains of 451 amino acids and two light chains of 213 amino acids
Catalog No. 50-271-8975 Supplier ACROBiosystems Supplier No. RIBFY35C25UG
 Restricted Item
Add to Cart
Add to Cart
Rituxan is a genetically engineered chimeric murine/human monoclonal antibody directed against the CD20 antigen found on the surface of normal and malignant B lymphocytes. The antibody is an IgG1 kappa immunoglobulin containing murine light- and heavy-chain variable region sequences and human constant region sequences. Rituximab is composed of two heavy chains of 451 amino acids and two light chains of 213 amino acids