Learn More
BIOHIPPO Mouse RLN3 Microsample
SDP
Catalog No. 5000315835
Encompass
Mouse RLN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN3 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 156 25-10000 pg/mL with a sensitivity of 42 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse RLN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse RLN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse RLN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15835 Supplier BIOHIPPO Supplier No. BHE15210873
Mouse RLN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN3 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 156 25-10000 pg/mL with a sensitivity of 42 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse RLN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse RLN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse RLN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color