Learn More
Sino Biological Recombinant Mouse ROR1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 5A0666M02H100
Encompass_Preferred
A DNA sequence encoding the Mouse ROR1 (NP_038873.2) (Met1-Glu403) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6539