Learn More
Biomatik Corporation Mouse R-Spondin 2 (RSPO2) ELISA Kit
Supplier:  Biomatik Corporation EKU09329
Encompass_Preferred
Mouse R-Spondin 2 (RSPO2) ELISA Kit for detecting RSPO2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BFU0. Gene ID: 239405. Target Synonyms: Roof plate-specific spondin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5826