Learn More
Sino Biological Anti-Nogo Receptor/RTN4R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50106RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Nogo Receptor / RTN4R (rM Nogo Receptor / RTN4R; Catalog#50106-M08H; NP_075358.1; Met1-Ser447). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3459