Learn More
Biomatik Corporation Mouse Ryanodine Receptor 2, Cardiac (RYR2) ELISA Kit
Supplier:  Biomatik Corporation EKU12528
Encompass_Preferred
Mouse Ryanodine Receptor 2, Cardiac (RYR2) ELISA Kit for detecting RYR2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E9Q401. Gene ID: 20191. Target Synonyms: RYR-2; ARVD2; ARVC2; VTSIP; Arrhythmogenic Right Ventricular Dysplasia 2; Cardiac muscle ryanodine receptor-calcium release channel. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7951