Learn More
Sino Biological Mouse S100A1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK5026615
Encompass_Preferred
The Mouse S100A1 ELISA Pair Set is for the quantitative determination of Mouse S100A1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse S100A1 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse S100A1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse S100A1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse S100A1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85475