Learn More
BIOHIPPO Mouse S100A10 Microsample
SDP
Catalog No. 5000314183
Encompass
Mouse S100A10 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A10 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse S100A10 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse S100A10 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse S100A10 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14183 Supplier BIOHIPPO Supplier No. BHE15212253
Mouse S100A10 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A10 in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse S100A10 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse S100A10 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse S100A10 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color