Learn More
Sino Biological Mouse S100A11 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50227T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse S100A11 (rM S100A11; Catalog#50227-M07E; NP_058020.1; Met1-Ile98). S100A11 specific IgG was purified by Mouse S100A11 affinity chromatography.
Catalog No. 50-237-0645