Learn More
Sino Biological Anti-S100A4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50484T20100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse S100A4 (rM S100A4; Catalog#50484-M07E; NP_035441.1; Met1-Lys101). S100A4 specific IgG was purified by Mouse S100A4 affinity chromatography.
Catalog No. 50-255-3255